Skip to Content
Merck
All Photos(3)

Key Documents

429031

Sigma-Aldrich

4,7-Dihydroxy-1,10-phenanthroline

Dye content ≥30 %

Synonym(s):

1,10-Phenanthroline-4,7-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2O2
CAS Number:
Molecular Weight:
212.20
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

>85.0% (HPLC)

Quality Level

form

powder or chunks

composition

Dye content, ≥30%

mp

>300 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccnc2c1ccc3c(O)ccnc23

InChI

1S/C12H8N2O2/c15-9-3-5-13-11-7(9)1-2-8-10(16)4-6-14-12(8)11/h1-6H,(H,13,15)(H,14,16)

InChI key

SLIBCJURSADKPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service