Skip to Content
Merck
All Photos(1)

Key Documents

426393

Sigma-Aldrich

1-tert-Butyl-3-ethylcarbodiimide

99%

Synonym(s):

N-(Ethylcarbonimidoyl)-2-methyl-2-propanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CN=C=NC2H5
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

reaction suitability

reaction type: Coupling Reactions

refractive index

n20/D 1.432 (lit.)

bp

137-139 °C (lit.)

density

0.814 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

functional group

amine

SMILES string

CCN=C=NC(C)(C)C

InChI

1S/C7H14N2/c1-5-8-6-9-7(2,3)4/h5H2,1-4H3

InChI key

AXTNYCDVWRSOCU-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Application

Reactant for:
  • Guanylation of amines
  • Synthesis of tantalum dimethylamido diamidato and guanidinato complexes for CVD of nitride thin films
  • Preparation of titanium amidinate guanidinate alkylamide complex precursors for HfO2 films
  • Ketene cycloadditions
  • Hydroamination of carbodiimides using group-2-element amides as precatalysts
  • Synthesis of cobalt amidinato derivative complexes for studies involving thermal stability, vapro pressure, vaporization rate and thermal decomposition properties

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service