Skip to Content
Merck
All Photos(2)

Key Documents

426008

Sigma-Aldrich

Carbofuran

98%

Synonym(s):

2,3-Dihydro-2,2-dimethyl-7-benzofuranol N-methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO3
CAS Number:
Molecular Weight:
221.25
Beilstein:
1428746
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

150-153 °C (lit.)

SMILES string

CNC(=O)Oc1cccc2CC(C)(C)Oc12

InChI

1S/C12H15NO3/c1-12(2)7-8-5-4-6-9(10(8)16-12)15-11(14)13-3/h4-6H,7H2,1-3H3,(H,13,14)

InChI key

DUEPRVBVGDRKAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbofuran is an anticholinesterase carbamate, which is commonly used as a pesticide. Its literature data on toxicity have been reviewed. The effect from exposure to carbofuran has been investigated on acetylcholinesterase (AChE) activity in the brain and muscle of common carp (Cyprinus carpio). Degradation studies using ozone, UV radiation and advanced oxidation processes (AOPs) and by photocatalytic degradation have been investigated. Ultra high performance liquid chromatography (UHPLC) method to determine the carbofuran in water samples have been developed with satisfactory results.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Degradation of carbofuran by using ozone, UV radiation and advanced oxidation processes.
Benitez FJ, et al.
Journal of Hazardous Materials, 89(1), 51-65 (2002)
Photocatalytic degradation of carbofuran using semiconductor oxides.
Mahalakshmi M, et al.
Journal of Hazardous Materials, 143(1), 240-245 (2007)
Li A Lu et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 47(6), 553-561 (2012-04-13)
Carbofuran, one of the most toxic and biorefractory carbamate compounds, is widely used in insecticides in Taiwan (9-18% of total insecticides production per year). In the present study, a central composite design experiment was used to study the effect of
Luís Pedro de Melo Plese et al.
Chemosphere, 60(2), 149-156 (2005-05-26)
The objectives of this work were estimate the reaction rates of hydrolysis of carbosulfan to carbofuran and subsequent degradation of this last compound in irrigated rice fields, and the respective half life, in aquatic environment and soil solution, by mean
Luz E Vera-Avila et al.
Talanta, 88, 553-560 (2012-01-24)
A selective and simple analytical method for the trace level determination of carbofuran in complex environmental and biological samples was developed based on immunoaffinity extraction (IAE) followed by on-line preconcentration and HPLC/UV analysis of the purified extract. The immunosorbent for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service