Skip to Content
Merck
All Photos(1)

Key Documents

423955

Sigma-Aldrich

Methyl 2-methylglycidate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.418 (lit.)

bp

62-65 °C/32 mmHg (lit.)

density

1.097 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

COC(=O)C1(C)CO1

InChI

1S/C5H8O3/c1-5(3-8-5)4(6)7-2/h3H2,1-2H3

InChI key

OSYXXQUUGMLSGE-UHFFFAOYSA-N

General description

Methyl 2-methylglycidate (methyl 2-methyloxirane-2-carboxylate) is an ester derived epoxide. It is reported to be formed by the reaction of methyl pyruvate with diazomethane. It is formed as an intermediate in the synthesis of 1,2,4-oxadiazole derivatives.

Application

Methyl 2-methylglycidate may be used in the synthesis of:
  • 2-methyl-2-hydroxy-1-propanol
  • 2,3-dihydroxy-2-methyl propionic acid
  • methyl 2-hydroxy-3-((4-methoxyphenyl)sulfonyl)-2-methylpropanoate

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gopal L Khatik et al.
Bioorganic & medicinal chemistry letters, 22(5), 1912-1916 (2012-02-14)
Sulfide and sulfonyl derivatives of 1,2,4-oxadiazoles were synthesized and screened by MTT assay on the prostate cancer cells, DU-145. Six compounds were identified as potential anti-prostate cancer agents with IC(50) values ranging from 0.5 to 5.1μM. These compounds exhibited good
DABSO-Based, Three-Component, One-Pot Sulfone Synthesis.
Deeming AS, et al.
Organic Letters, 16(1), 150-153 (2013)
β-Keto phosphonic esters.
Arbuzov BA, et al.
Russian Chemical Bulletin, 12(4), 604-610 (1963)
R J Steffan et al.
Applied and environmental microbiology, 63(11), 4216-4222 (1997-11-15)
Several propane-oxidizing bacteria were tested for their ability to degrade gasoline oxygenates, including methyl tert-butyl ether (MTBE), ethyl tert-butyl ether (ETBE), and tert-amyl methyl ether (TAME). Both a laboratory strain and natural isolates were able to degrade each compound after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service