Skip to Content
Merck
All Photos(1)

Key Documents

421286

Sigma-Aldrich

MERPOL® A surfactant

Synonym(s):

Nonionic Surfactant MERPOL

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

Quality Level

refractive index

n20/D 1.453 (lit.)

surface tension

24 dyn/cm, 25 °C, 0.05 wt. %

bp

200 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

HLB

6

Application

Nonionic, ethylene oxide condensate. Low foaming wetting agent.

Legal Information

MERPOL is a registered trademark of Stepan Co.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service