Skip to Content
Merck
All Photos(2)

Key Documents

418978

Sigma-Aldrich

(DHQ)2Pyr

97%

Synonym(s):

Hydroquinine 2,5-diphenyl-4,6-pyrimidinediyl diether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H60N6O4
CAS Number:
Molecular Weight:
881.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D +455°, c = 1.2 in methanol

impurities

<3% ethyl acetate

mp

245-248 °C (lit.)

functional group

phenyl

SMILES string

CC[C@H]1CN2CCC1C[C@@H]2[C@H](Oc3nc(nc(O[C@@H]([C@@H]4CC5CC[N@H]4C[C@H]5CC)c6ccnc7ccc(OC)cc67)c3-c8ccccc8)-c9ccccc9)c%10ccnc%11ccc(OC)cc%10%11

InChI

1S/C56H60N6O4/c1-5-35-33-61-27-23-39(35)29-49(61)52(43-21-25-57-47-19-17-41(63-3)31-45(43)47)65-55-51(37-13-9-7-10-14-37)56(60-54(59-55)38-15-11-8-12-16-38)66-53(50-30-40-24-28-62(50)34-36(40)6-2)44-22-26-58-48-20-18-42(64-4)32-46(44)48/h7-22,25-26,31-32,35-36,39-40,49-50,52-53H,5-6,23-24,27-30,33-34H2,1-4H3/t35-,36?,39?,40+,49?,50?,52+,53+/m0/s1

InChI key

SWKRDCRSJPRVNF-RMMANSCUSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Sold under license from Rhodia Pharma Solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service