Skip to Content
Merck
All Photos(1)

Key Documents

418293

Sigma-Aldrich

Sodium thiophenolate

technical grade, 90%

Synonym(s):

Benzenethiol sodium salt, Sodium thiophenoxide, Thiophenol sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SNa
CAS Number:
Molecular Weight:
132.16
Beilstein:
3597302
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

powder

mp

>300 °C (lit.)

SMILES string

[Na]Sc1ccccc1

InChI

1S/C6H6S.Na/c7-6-4-2-1-3-5-6;/h1-5,7H;/q;+1/p-1

InChI key

RZWQDAUIUBVCDD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium thiophenolate can be prepared from the reaction of sodium and thiophenol in diethyl ether.

Application

Sodium thiophenolate has been used for the synthesis of MCoTI-I and MCoTI-II cyclotides, which are naturally-occurring cyclic cystine-knot microprotein trypsin inhibitors. It may be employed in the following studies:
  • As probe for the immunoassay and for the detection of label-free protein by surface-enhanced Raman scattering (SERS).
  • Preparation of new cyclometalated 6-phenyl-4-(p-R-phenyl)-2,2′-bipyridyl (C--N--N)Pt(II) thiophenolate complexes.
  • Synthesis of 1,3,5,7,9-pentakis(4-methoxyphenylthio)corannulene, 1,3,5,7,9-pentakis(2-naphthylthio)corannulene and 1,3,6,8-tetrakis(4-methoxyphenylthio)corannulene.
  • Synthesis of Et4N+ salts of homoleptic arylthiolate Ti(IV) complex, [Ti(SPh)6]2-.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mizyed et al.
Journal of the American Chemical Society, 123(51), 12770-12774 (2001-12-26)
1,3,5,7,9-Pentakis(4-methoxyphenylthio)corannulene (3), 1,3,5,7,9-pentakis(2-naphthylthio)corannulene (4), and 1,3,6,8-tetrakis(4-methoxyphenylthio)corannulene (5b) have been synthesized by chlorination of corannulene with ICl in CH(2)Cl(2) at 25 degrees C and subsequent nucleophilic aromatic substitution with the appropriate sodium thiophenolate in DMEU at 25 degrees C. (1)H NMR
Panumart Thongyoo et al.
Organic & biomolecular chemistry, 6(8), 1462-1470 (2008-04-04)
The naturally-occurring cyclic cystine-knot microprotein trypsin inhibitors MCoTI-I and MCoTI-II have been synthesised using both thia-zip native chemical ligation and a biomimetic strategy featuring chemoenzymatic cyclisation by an immobilised protease. Engineered analogues have been produced containing a range of substitutions
Jacob Schneider et al.
Inorganic chemistry, 48(4), 1498-1506 (2009-01-15)
Three new cyclometalated 6-phenyl-4-(p-R-phenyl)-2,2'-bipyridyl (C--N--N) Pt(II) thiophenolate complexes (R = Me (2a), COOMe (2b), and P(O)(OEt)(2) (2c)) have been synthesized and studied. The new C--N--N ligands L2 (R = COOMe) and L3 (R = P(O)(OEt)(2)) undergo cyclometalation with a Pt(II)
Xuefang Gu et al.
Analytical and bioanalytical chemistry, 406(7), 1885-1894 (2014-03-01)
A simple and sensitive method, based on surface-enhanced Raman scattering (SERS), for immunoassay and label-free protein detection is reported. A series of bowl-shaped silver cavity arrays were fabricated by electrodeposition using a self-assembled polystyrene spheres template. The reflection spectra of
The synthesis and structural characterization of [Et4N]2 [Ti(SPh)6] and trimeric [Ti3O(SPh)3Cl4 (CH3 CN)5]?CH3 CN?(C2 H5)2O complexes.
Kim JT, et al.
Polyhedron, 19(9), 1139-1143 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service