Skip to Content
Merck
All Photos(1)

Key Documents

418072

Sigma-Aldrich

Carbon-12C monoxide

99.95 atom % 12C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12CO
CAS Number:
Molecular Weight:
28.00
EC Number:
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

description

13C-depleted

Quality Level

isotopic purity

99.95 atom % 12C

Assay

99% (CP)

bp

-191.5 °C (lit.)

mp

-205 °C (lit.)

mass shift

depleted

SMILES string

[12C-]#[O+]

InChI

1S/CO/c1-2/i1+0

InChI key

UGFAIRIUMAVXCW-IGMARMGPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard products are packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressures at 21ºC are 320 psig for 10 L and 850 psig for 25 L. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Gas 1A - Press. Gas Compr. Gas - Repr. 1A - STOT RE 1 Inhalation

Target Organs

Heart

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farhad Peerani et al.
Inflammatory bowel diseases, 21(8), 1817-1824 (2015-06-19)
Pulmonary involvement is a recognized extraintestinal complication of inflammatory bowel disease and is associated with airway inflammation. Changes in pulmonary function were previously described as being subclinical. The purpose of this study was to compare pulmonary findings in a large
Elizabeth R Aston et al.
Drug and alcohol dependence, 152, 32-38 (2015-05-24)
A valid measure of the relative economic value of marijuana is needed to characterize individual variation in the drug's reinforcing value and inform evolving national marijuana policy. Relative drug value (demand) can be measured via purchase tasks, and demand for
T A Duong et al.
The British journal of dermatology, 172(2), 400-405 (2014-12-17)
Acute-stage specific bronchial epithelial detachment has been described in 27% of patients with Stevens-Johnson syndrome (SJS) and toxic epidermal necrolysis (TEN). To assess the pulmonary function of patients with SJS/TEN after remission. Analysis of pulmonary function tests (PFTs) performed during
Marc A Judson et al.
American journal of respiratory and critical care medicine, 191(7), 786-795 (2015-01-17)
Patient-reported outcome (PRO) measures have been developed to measure symptoms and other aspects of health-related quality of life. The Sarcoidosis Assessment Tool (SAT), a sarcoidosis-specific PRO, was administered in a lung and skin sarcoidosis treatment trial. We explored SAT performance
Takashi Kurita et al.
Rheumatology (Oxford, England), 54(1), 39-44 (2014-04-26)
Interstitial lung diseases (ILDs) complicated with PM or DM are frequently aggressive and refractory to treatment. Recently some reports have suggested the potential benefit of tacrolimus for severe ILD complicated with PM/DM. However, little evidence has yet shown the efficacy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service