Skip to Content
Merck
All Photos(1)

Key Documents

415774

Sigma-Aldrich

5′-Chloro-2′-hydroxy-3′-nitroacetophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(OH)(NO2)COCH3
CAS Number:
Molecular Weight:
215.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

132-135 °C (lit.)

functional group

chloro
ketone
nitro

SMILES string

CC(=O)c1cc(Cl)cc(c1O)[N+]([O-])=O

InChI

1S/C8H6ClNO4/c1-4(11)6-2-5(9)3-7(8(6)12)10(13)14/h2-3,12H,1H3

InChI key

IUNBIQBAYUBIFD-UHFFFAOYSA-N

General description

5′-Chloro-2′-hydroxy-3′-nitroacetophenone (5-chloro-3-nitro-2-hydroxyacetophenone) is a nitro derivative of ortho-hydroxy aryl ketone. Its crystal structure has been studied by X-ray diffraction. It forms complex with 2-aminobenzoic acid (anthranilic acid), in which the nitro group is turned by approximately 40°C.

Application

5′-Chloro-2′-hydroxy-3′-nitroacetophenone may be used for the synthesis of 6-chloro-8-nitro-4-oxo-4H-chromene-3-carbaldehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase transition and intramolecular hydrogen bonding in nitro derivatives of ortho-hydroxy acetophenones
Filarowski, A., et al.
Journal of Molecular Structure, 785(1), 7-13 (2006)
Yoshinobu Ishikawa
Acta crystallographica. Section E, Structure reports online, 70(Pt 5), o547-o547 (2014-05-27)
In the title compound, C10H4ClNO5, the non-H atoms of the 6-chloro-chromone unit are coplanar (r.m.s. deviation = 0.017 Å) with the largest deviation from the mean plane [0.031 (2) Å] being found for the C=O C atom. The nitro group (NO2) is inclined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service