Skip to Content
Merck
All Photos(1)

Key Documents

414638

Sigma-Aldrich

mono-tert-Butyl malonate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCOCH2COOH
CAS Number:
Molecular Weight:
160.17
Beilstein:
1765457
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

impurities

5% methanesulfonyl chloride

refractive index

n20/D 1.426 (lit.)

bp

90 °C/2 mmHg (lit.)

mp

19-20 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC(=O)CC(O)=O

InChI

1S/C7H12O4/c1-7(2,3)11-6(10)4-5(8)9/h4H2,1-3H3,(H,8,9)

InChI key

NGGGZUAEOKRHMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mono-tert-Butyl malonates is an ester. It is reported to be an aminoacylase inhibitor. Preparation of mono-tert-Butyl malonates has been described.

Application

Mono-tert-Butyl malonates may be used in the preparation of the following:
  • dendritic precursor to asymmetric methanofullerenes
  • hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid
  • hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of high-aspect-ratio helical nanorods via chiral self-assembly of fullerodendrimers.
Hilmer AJ, et al.
The Journal of Physical Chemistry Letters, 5(5), 929-934 (2014)
Helen S Toogood et al.
Extremophiles : life under extreme conditions, 6(2), 111-122 (2002-05-16)
A thermostable L-aminoacylase from Thermococcus litoralis was cloned, sequenced, and overexpressed in Escherichia coli. The enzyme is a homotetramer of 43 kDa monomers and has an 82% sequence identity to an aminoacylase from Pyrococcus horikoshii and 45% sequence identity to
Facile Preparation and Purification of Mono tert-Butyl Malonate.
Tararov VI, et al.
Synthetic Communications, 36(2), 187-191 (2006)
Jae Koo Lee et al.
Journal of agricultural and food chemistry, 51(13), 3695-3703 (2003-06-12)
A competitive indirect enzyme-linked immunosorbent assay (ciELISA) for the organophosphorus insecticide acephate, O,S-dimethyl acetylphosphoramidothioate, was developed using a polyclonal antibody. Five different haptens mimicking the analyte were synthesized and conjugated with the carrier proteins bovine serum albumin (BSA) and keyhole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service