Skip to Content
Merck
All Photos(1)

Key Documents

412813

Sigma-Aldrich

N,N-Diglycidyl-4-glycidyloxyaniline

Synonym(s):

4-(Diglycidylamino)phenyl glycidyl ether, 4-Glycidyloxy-N ,N -diglycidylaniline, N ,N ,o -Triglycidyl-p -aminophenol, p -(Diglycidylamino)phenyl glycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO4
CAS Number:
Molecular Weight:
277.32
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.567 (lit.)

density

1.22 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1OC1COc2ccc(cc2)N(CC3CO3)CC4CO4

InChI

1S/C15H19NO4/c1-3-12(17-9-15-10-20-15)4-2-11(1)16(5-13-7-18-13)6-14-8-19-14/h1-4,13-15H,5-10H2

InChI key

AHIPJALLQVEEQF-UHFFFAOYSA-N

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Muta. 2 - Skin Sens. 1 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giovanni Fortunato et al.
Polymers, 11(6) (2019-06-09)
Efficient and comprehensive recycling of fiber-reinforced thermosets is particularly challenging, since the irreversible degradation of the matrix component is necessary in order to separate the fiber component in high purity. In this work, a new approach to fully recyclable thermoset
Michael Giebler et al.
Polymers, 12(5) (2020-05-21)
Epoxy-anhydride vitrimers are covalent adaptable networks, which undergo associative bond exchange reactions at elevated temperature. Their service temperature is influenced by the glass transition temperature (Tg) as well as the topology freezing transition temperature (Tv), at which the covalent bond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service