Skip to Content
Merck
All Photos(2)

Key Documents

412325

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)

average Mn ~8,400

Synonym(s):

Pluronic® F-68 LF Pastille, PEG-PPG-PEG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.3 mmHg ( 20 °C)

form

beads

mol wt

average Mn ~8,400

composition

PEG, 80 wt. %

surface tension

50 dyn/cm, 25 °C, 0.1 wt. % in H2O

viscosity

1,000 cP(77 °C, Brookfield)(lit.)

transition temp

cloud point >100 °C (1 wt. % aqueous solution)
Tm 52 °C

density

1.06 g/mL at 25 °C

HLB

>24.0

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Surface active material whose properties are dependent on the hydrophilic (EO)/hydrophobic (PO) ratio. High EO content gives better o/w stabilizers.

Physical form

Terminal primary dihydroxy-functional oligomer.

Legal Information

Pluronic is a registered trademark of BASF

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean P Cleary et al.
The Analyst, 145(13), 4688-4697 (2020-05-28)
Native electrospray mass spectrometry is a powerful method for determining the native stoichiometry of many polydisperse multi-subunit biological complexes, including multi-subunit protein complexes and lipid-bound transmembrane proteins. However, when polydispersity results from incorporation of multiple copies of two or more
Neeta Jain-Gupta et al.
FEMS immunology and medical microbiology, 66(3), 436-444 (2012-11-21)
Brucellosis is the most common zoonotic disease worldwide, and there is no vaccine for human use. Brucella melitensis Rev1, a live attenuated strain, is the commercial vaccine for small ruminants to prevent B. melitensis infections but has been associated with abortions
Wanhua Feng et al.
Colloids and surfaces. B, Biointerfaces, 105, 43-50 (2013-01-29)
The interaction between bacteria and surfaces is central to many environmental, industrial and medical applications. Surfactants are commonly used in these applications and can potentially influence the bacterium/surface interaction. The effect of surfactants upon bacterial cell surface thermodynamic properties was
Yanbin Guan et al.
Archives of pharmacal research, 34(10), 1719-1728 (2011-11-15)
The aim of the present study was to evaluate the effect of pluronic P123 (P123) and pluronic F127 (F127) on intestinal P-glycoprotein (P-gp) and cytochrome P450 3A using the specific substrates rhodamine-123 (R-123) and midazolam, respectively. Caco-2 cells and everted
Mi Mi Wan et al.
Journal of colloid and interface science, 377(1), 497-503 (2012-04-18)
New concept on the promotion of immobilization and catalytic activity of enzyme on mesoporous silica through template micelles is proposed and realized in this paper. Proper P123 templates are controllable retained in the as-synthesized SBA-15, not only to anchor the

Articles

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service