Skip to Content
Merck
All Photos(2)

Key Documents

411914

Sigma-Aldrich

3-Nitrophthalonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CN)2
CAS Number:
Molecular Weight:
173.13
Beilstein:
2263686
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

99%

form

solid

mp

162-165 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-][N+](=O)c1cccc(C#N)c1C#N

InChI

1S/C8H3N3O2/c9-4-6-2-1-3-8(11(12)13)7(6)5-10/h1-3H

InChI key

UZJZIZFCQFZDHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Taner Arslan et al.
Journal of fluorescence, 30(2), 365-373 (2020-02-23)
In present work, a new substituted phthalonitrile derivative was prepared by the nucleophilic displacement reaction and then highly soluble zinc phthalocyanine (ZnPc) with four peripheral 1-hydroxyhexan-3-ylthio groups was synthesized by cyclotetramerization and characterized by FTIR, 1H and 13C NMRs spectroscopies
Armağan Günsel et al.
Dalton transactions (Cambridge, England : 2003), 48(39), 14839-14852 (2019-09-26)
This study describes the synthesis and characterization of a non-peripherally tetra-substituted copper(ii) phthalocyanine bearing 4-(trifluoromethoxy)phenol groups. Some spectroscopic techniques such as FT-IR, 1H-NMR, 13C-NMR, MALDI-TOF, SEM and UV-vis were used to characterize the compounds. The novel molecule of 3-(4-(trifluoromethoxy) phenoxy)
G S Amitha et al.
Journal of fluorescence, 29(1), 279-291 (2019-01-10)
A novel 4,4'-fluoresceinoxy bisphthalonitrile FPN is synthesized from fluorescein and 4-nitrophthalonitrile by aromatic nucleophilic ipso nitro substitution reaction. The structure of FPN constitutes phthalonitrile-fluorescein-phthalonitrile, acceptor-donor-acceptor, A-D-A form and the solvatochromic study of newly synthesized compound FPN was done in hexane
Min Wang et al.
Nature communications, 10(1), 3602-3602 (2019-08-11)
Molecular catalysts that combine high product selectivity and high current density for CO2 electrochemical reduction to CO or other chemical feedstocks are urgently needed. While earth-abundant metal-based molecular electrocatalysts with high selectivity for CO2 to CO conversion are known, they

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service