409189
Glycerol ethoxylate-co-propoxylate triol
average Mn ~2,600
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
HO(C3H6O)x(CH2CH2O)yCH[CH2(OCH2CH2)y(OC3H6)xOH]2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
solid
mol wt
average Mn ~2,600
refractive index
n20/D 1.462 (lit.)
density
1.063 g/mL at 25 °C (lit.)
SMILES string
OCCO.OCCCO.OCC(O)CO
InChI
1S/C3H8O3.C3H8O2.C2H6O2/c4-1-3(6)2-5;4-2-1-3-5;3-1-2-4/h3-6H,1-2H2;4-5H,1-3H2;3-4H,1-2H2
InChI key
ZTVLGYSTCFTIBJ-UHFFFAOYSA-N
Related Categories
Application
Glycerol ethoxylate-co-propoxylate triol is a cross-linker that can be used as a UV curable binder for polyurethane acrylate. It has a low viscosity that makes it useful in the preparation of inks for ink-jet printing and the UV based drying of these pigments.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effect of Poly (ethylene glycol) dimethyl ether Plasticizer on Ionic Conductivity of Cross-Linked Poly [siloxane-g-oligo (ethylene oxide)] Solid Polymer Electrolytes
Kang Y, et al.
Macromolecular Research, 12(5), 431-436 (2004)
Liyuan Hou et al.
Protoplasma, 251(1), 191-199 (2013-08-15)
We tested for chemical reagents that would be useful in preparing a large number of vital single cells from colonial Botryococcus braunii B-race, variety Showa. Among the 18 reagents assayed, glycerol and erythritol showed the highest potency for releasing single
Synthesis and characterization of aqueous UV-curable binder for ink preparation in ink-jet printing and pigment dyeing of fabrics
El-Molla MM
Indian journal of fibre & textile research, 32(5), 105-113 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service