Skip to Content
Merck
All Photos(1)

Key Documents

408999

Sigma-Aldrich

1,3-Butanediol dimethacrylate

contains 150-250 ppm MEHQ as inhibitor, 95%

Synonym(s):

1,3-Butylene dimethacrylate, 1,3-Butyleneglycol dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2CH(CH3)O2CC(CH3)=CH2
CAS Number:
Molecular Weight:
226.27
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

liquid

contains

150-250 ppm MEHQ as inhibitor

refractive index

n20/D 1.452 (lit.)

bp

290 °C (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

CC(CCOC(=O)C(C)=C)OC(=O)C(C)=C

InChI

1S/C12H18O4/c1-8(2)11(13)15-7-6-10(5)16-12(14)9(3)4/h10H,1,3,6-7H2,2,4-5H3

InChI key

VDYWHVQKENANGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service