408794
Lithium methoxide solution
1.0 M in methanol
Synonym(s):
Lithium methylate
About This Item
Recommended Products
form
liquid
Quality Level
reaction suitability
core: lithium
concentration
1.0 M in methanol
bp
64.6 °C
density
0.817 g/mL at 25 °C
SMILES string
[Li+].C[O-]
InChI
1S/CH3O.Li/c1-2;/h1H3;/q-1;+1
InChI key
JILPJDVXYVTZDQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B - STOT SE 1
Target Organs
Eyes
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 2
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service