Skip to Content
Merck
All Photos(1)

Key Documents

405493

Sigma-Aldrich

Poly(Bisphenol A-co-epichlorohydrin), glycidyl end-capped

average Mn ~377

Synonym(s):

Bisphenol A diglycidyl ether-bisphenol A copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

mol wt

average Mn ~377

refractive index

n20/D 1.573

viscosity

100-150 poise(25 °C)

density

1.16 g/mL at 25 °C

SMILES string

ClCC4OC4.O3C(C3)CO.Oc1ccc(cc1)C(C)(C)c2ccc(cc2)O.OC(CO)CO

InChI

1S/C15H16O2.C3H5ClO.C3H8O3.C3H6O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;4-1-3-2-5-3;4-1-3(6)2-5;4-1-3-2-5-3/h3-10,16-17H,1-2H3;3H,1-2H2;3-6H,1-2H2;3-4H,1-2H2

InChI key

YCDGZAHDCFWTHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Martin-Gallego et al.
Polymers, 11(5) (2019-05-06)
Owing to their high strength and stiffness, thermal and environmental stability, lower shrinkage, and water resistance, epoxy resins have been the preferred matrix for the development of syntactic foams using hollow glass microspheres. Although these foams are exploited in multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service