Skip to Content
Merck
All Photos(3)

Key Documents

402583

Sigma-Aldrich

Zinc

shot, <12 mm, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Quality Level

Assay

99.99% trace metals basis

form

shot

resistivity

5.8 μΩ-cm, 20°C

particle size

<12 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A viewpoint about the treatment of Wilson's disease.
Abdul Qayyum Rana et al.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques, 40(4), 612-614 (2013-06-22)
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking
Bikha Ram Devrajani et al.
JPMA. The Journal of the Pakistan Medical Association, 63(3), 369-373 (2013-08-07)
To determine the serum zinc level in patients with pneumonia. The descriptive cross-sectional study was conducted at the Liaquat University Hospital, Hyderabad, Pakistan, from September 2008 to February 2009. All patients above 12 years of age and of either gender
Lena Martin et al.
Archives of animal nutrition, 67(3), 248-261 (2013-06-08)
This study aimed at investigating the effect of dietary zinc on performance, jejunal brush border membrane enzyme activities and mRNA levels of enzymes and two zinc transporters in piglets. A total of 126 piglets were weaned at 26 ±1 days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service