Skip to Content
Merck
All Photos(1)

Key Documents

401455

Sigma-Aldrich

Ethyl 4,4,4-trifluoro-2-butynoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C≡CCO2C2H5
CAS Number:
Molecular Weight:
166.10
Beilstein:
3539414
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.350 (lit.)

bp

96-98 °C (lit.)

density

1.162 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

CCOC(=O)C#CC(F)(F)F

InChI

1S/C6H5F3O2/c1-2-11-5(10)3-4-6(7,8)9/h2H2,1H3

InChI key

SFDRHPQGYUYYNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 4,4,4-trifluoro-2-butynoate is an unsymmetrical internal alkyne.

Application

Ethyl 4,4,4-trifluoro-2-butynoate has been used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes.

It may be used in the synthesis of the following compounds :
  • (2S,4aR,6aR,7R,9S,10aS,10bR)-methyl 9-acetoxy-2-(5-(ethoxycarbonyl)-6-(trifluoromethyl)-7-oxabicyclo[2.2.1]hepta-2,5-dien-2-yl)-6a,10b-dimethyl-4,10-dioxododecahydro-1H-benzo[f]isochromene-7-carboxylate
  • (2S,4aR,6aR,7R,9S,10aS,10bR)-methyl 9-acetoxy-2-(6-(ethoxycarbonyl)-5-(trifluoromethyl)-7-oxabicyclo[2.2.1]hepta-2,5-dien-2-yl)-6a,10b-dimethyl-4,10-dioxododecahydro-1Hbenzo[f]isochromene-7-carboxylate

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactivity and Regioselectivity of Insertion of Unsaturated Molecules into M- C (M= Ir, Rh) Bonds of Cyclometalated Complexes.
Li L, et al.
Organometallics, 29(20), 4593-4605 (2010)
Facile access to stereodefined dienoates and cyclopropylenoates containing a trifluoromethyl group.
Wang P-A, et al.
Journal of Fluorine Chemistry, 124(1), 93-97 (2003)
Anthony Lozama et al.
Journal of natural products, 74(4), 718-726 (2011-02-23)
As part of our continuing efforts toward more fully understanding the structure-activity relationships of the neoclerodane diterpene salvinorin A, we report the synthesis and biological characterization of unique cycloadducts through [4+2] Diels-Alder cycloaddition. Microwave-assisted methods were developed and successfully employed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service