Skip to Content
Merck
All Photos(2)

Key Documents

397210

Sigma-Aldrich

2-Amino-5-chloro-2′-fluorobenzophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Cl)C(O)C6H4F
CAS Number:
Molecular Weight:
249.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

95-98 °C (lit.)

functional group

chloro
fluoro
ketone

SMILES string

Nc1ccc(Cl)cc1C(=O)c2ccccc2F

InChI

1S/C13H9ClFNO/c14-8-5-6-12(16)10(7-8)13(17)9-3-1-2-4-11(9)15/h1-7H,16H2

InChI key

GTGMXPIQRQSORU-UHFFFAOYSA-N

Application

2-Amino-5-chloro-2′-fluorobenzophenone is suitable electrophilic coupling reagent for the determination of iron(III) in environmental waters, soils and industrial effluent samples. It may be used in the synthesis of following:
  • 2,4-disubstituted quinolones, via Meyer-Schuster rearrangement
  • 6-chloro-2-methyl-4-(2-fluorophenyl) quinazoline
  • N-desalkylflurazepam
  • benzotriazepines
  • diazepam-related benzodiazepines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ionic liquid-an efficient recycalable system for the synthesis of 2, 4-disubstituted quinolines via Meyer-Schuster rearrangement.
Saqrma R and Prajapati D.
Synlett, 19, 3001-3005 (2008)
Bhawana Sati et al.
Acta pharmaceutica (Zagreb, Croatia), 63(3), 385-396 (2013-10-25)
During the manufacture of bulk drug midazolam various impurities arised that can be the related products or degradation products. Structures of eight impurities that can arise during the manufacture of bulk drug midazolam were proposed. In the present work, synthesis
J. Chem. Res. Synop., 2-2 (1991)
Journal of the American Chemical Society, 112, 3969-3969 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service