Skip to Content
Merck
All Photos(1)

Key Documents

397180

Sigma-Aldrich

Dimethyl trithiocarbonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3S)2CS
CAS Number:
Molecular Weight:
138.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.675 (lit.)

bp

101-102 °C/12 mmHg (lit.)

mp

−3 °C (lit.)

density

1.254 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

CSC(=S)SC

InChI

1S/C3H6S3/c1-5-3(4)6-2/h1-2H3

InChI key

IQWMXKTYXNMSLC-UHFFFAOYSA-N

Application

Dimethyl trithiocarbonate may be used in the following studies:
  • Preparation of methyl-β,β′-dicarbonyldithiocarboxylate derivatives.
  • Generation of tris(organothiyl)methyl radicals and these radicals were evaluated using EPR spectroscopy.
  • Preparation of β-oxodithiocarboxylates.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An electron paramagnetic resonance study of free-radical additions to trithiocarbonates and of the formation and destruction of tetrathiafulvalene by free-radical processes.
Forrest D and Ingold KU.
Journal of the American Chemical Society, 100(12), 3868-3873 (1978)
Gas-Phase Thermolysis, 14. On the Isomerization of Dimethyl Carbonate and Its Mono-, Di-, and Trithio Analogs.
Egsgaard H, et al.
Chemische Berichte, 124(5), 1265-1270 (1999)
A New Application of Dimethyl Trithiocarbonate. Methylthiothiocarbonylation of 2, 4-Pentanedione and Some β-Oxoesters.
Oliva A, et al.
Synthetic Communications, 28(18), 3381-3386 (1998)
A facile method for the synthesis of substituted 2-ylidene-1, 3-oxathioles from acetophenones.
Samuel R, et al.
Tetrahedron Letters, 48(47), 8376-8378 (2007)
Dirk Stueber et al.
Solid state nuclear magnetic resonance, 22(4), 439-457 (2003-01-24)
The 13C chemical shift tensor principal values for the trigonal carbonate and thiocarbonate carbon atoms in the dialkyl carbonates, dimethyl carbonate, ethylene carbonate, and diphenyl carbonate, and in the trithiocarbonates, ethylene trithiocarbonate and dimethyl trithiocarbonate, respectively, were measured in various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service