All Photos(1)
About This Item
Linear Formula:
Br(CH2)6CO2C2H5
CAS Number:
Molecular Weight:
237.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.459 (lit.)
bp
112 °C/5 mmHg (lit.)
mp
29 °C (lit.)
density
1.217 g/mL at 25 °C (lit.)
functional group
bromo
ester
SMILES string
CCOC(=O)CCCCCCBr
InChI
1S/C9H17BrO2/c1-2-12-9(11)7-5-3-4-6-8-10/h2-8H2,1H3
InChI key
OOBFNDGMAGSNKA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Ethyl 7-bromoheptanoate is a bromoester. It is used as an alkylating agent for organic compounds. It participates in the synthesis of ω-chain shortened prostaglandins.
Application
Ethyl 7-bromoheptanoate may be used in the following studies:
- As a starting material in the synthesis of benzyl 7-iodoheptanoate.
- As one of the reactant in the synthesis of diethyl 7,7′-(2,5-diiodo-1,4-phenylene)bis(oxy)diheptanoate.
- As one of the reagent in the synthesis of CarnDOD-7C, a carnitine-derived nitrone.
- As an alkylating agent in the alkylation of ethyl 3-oxoglutarate to form 2-(6-ethoxycarbonylhexyl)-3-oxoglutarate.
- Synthesis of 1-substituted cyclopentadiene by alkylating lithium cyclopentadienide.5
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Siham El Fangour et al.
Age (Dordrecht, Netherlands), 31(4), 269-276 (2009-05-30)
Oxidative damage from reactive oxygen species (ROS) and the carbon-centred radicals arising from them is important to the process of aging, and age-related diseases are generally caused, exacerbated or mediated by oxidative stress. Nitrones can act as spin traps to
Design Principle of Conjugated Polyelectrolytes to Make Them Water-Soluble and Highly Emissive.
Lee K, et al.
Advances in Functional Materials, 22(5), 1076-1086 (2012)
Synthesis of a Prostaglandin Intermediate and Synthesis of Dihydrojasmone and Methyl Dihydrojasmonate.
Naoshima Y, et al.
Agricultural and Biological Chemistry, 43(8), 1765-1768 (1979)
David J Hart
Organic Synthesis via Examination of Selected Natural Products, 99-99 null
Cinzia B Botta et al.
Journal of medicinal chemistry, 54(7), 2165-2182 (2011-03-23)
Several oxime containing molecules, characterized by a SAHA-like structure, were explored to select a potentially new biasing binding element for the zinc in HDAC catalytic site. All compounds were evaluated for their in vitro inhibitory activity against the 11 human
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service