Skip to Content
Merck
All Photos(1)

Key Documents

39030

Sigma-Aldrich

N,N-Dimethylethylenediamine

≥98.0% (GC)

Synonym(s):

2-(Dimethylamino)ethylamine, DMEN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2NH2
CAS Number:
Molecular Weight:
88.15
Beilstein:
605279
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.426 (lit.)
n20/D 1.427

bp

104-106 °C (lit.)

density

0.807 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CN(C)CCN

InChI

1S/C4H12N2/c1-6(2)4-3-5/h3-5H2,1-2H3

InChI key

DILRJUIACXKSQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylethylenediamine is a bidentate ligand that can be used:
  • To synthesize Schiff base ligands to prepare their zinc alkyl and zinc alkoxide complexes that initiate ring-opening polymerization of lactides.
  • As a ligand to prepare copper azido polymers.
  • In the synthesis of naphthalimides such as amonafide, an antitumor agent.
  • As the receptor in building fluorescent signaling systems to perturb the photoinduced intramolecular electron transfer (PET) process from the nitrogen lone-pair to the fluorophore.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.1 °F - closed cup

Flash Point(C)

11.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei Zhao et al.
Langmuir : the ACS journal of surfaces and colloids, 35(17), 5779-5786 (2019-01-24)
Nosocomial infections are a major problem in medical health care. To solve this problem, a series of antimicrobial waterborne paints were prepared by using antimicrobial hyperbranched (HB) emulsifiers. The HB-emulsifiers were prepared by polymerizing AB2 monomers obtained in a one-step
Intramolecular Aryne-Furan Cycloadditions for the Synthesis of Anticancer Naphthalimides.
Prevost S, et al.
The Journal of Organic Chemistry, 83(8), 4871-4881 (2018)
Synthesis, structures, and magnetic behavior of a series of copper (II) azide polymers of Cu4 building clusters and isolation of a new hemiaminal ether as the metal complex.
Mukherjee S, et al.
Inorganic Chemistry, 50(8), 3621-3631 (2011)
Ring-opening polymerization of lactides initiated by zinc alkoxides derived from NNO-tridentate ligands.
Chen H Y, et al.
Macromolecules, 39(11), 3745-3752 (2006)
Perturbation of the PET process in fluorophore? spacer? receptor systems through structural modification: Transition metal induced fluorescence enhancement and selectivity.
Bag B and Bharadwaj P K
The Journal of Physical Chemistry B, 109(10), 4377-4390 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service