Skip to Content
Merck
All Photos(1)

Key Documents

390151

Sigma-Aldrich

1,3-Bis(4-nitrophenyl)urea

97%

Synonym(s):

4′,4′′-Dinitrocarbanilide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2NC6H4NH)2CO
CAS Number:
Molecular Weight:
302.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

>300 °C (lit.)

functional group

amine
nitro

SMILES string

[O-][N+](=O)c1ccc(NC(=O)Nc2ccc(cc2)[N+]([O-])=O)cc1

InChI

1S/C13H10N4O5/c18-13(14-9-1-5-11(6-2-9)16(19)20)15-10-3-7-12(8-4-10)17(21)22/h1-8H,(H2,14,15,18)

InChI key

JEZZOKXIXNSKQD-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

standard

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Beier et al.
Journal of agricultural and food chemistry, 49(10), 4542-4552 (2001-10-16)
A cELISA was developed for the coccidiostat nicarbazin. On the basis of previous computer-assisted molecular modeling studies, p-nitrosuccinanilic acid (PNA-S) was selected as a hapten to produce antibodies to 4,4'-dinitrocarbanilide (DNC), the active component of the coccidiostat nicarbazin. Synthesis is
J E Jones et al.
Poultry science, 69(3), 378-387 (1990-03-01)
Two studies were conducted to determine the effects of anticoccidial agents on the production and reproduction of White Leghorns. In Experiment 1, nicarbazin (NCZ) was fed at 0, 20, 50, and 100 ppm. Hen-day egg production, egg weight, the egg-yolk
J A Tarbin et al.
Journal of chromatography, 613(2), 354-358 (1993-04-02)
A high-performance liquid chromatographic method for the determination of the dinitrocarbanilide component of the anti-coccidial drug nicarbazin has been developed. The drug was extracted from egg with acetonitrile. The extract was evaporated to dryness and taken up in water-acetonitrile (80:20
E F Rogers et al.
Science (New York, N.Y.), 222(4624), 630-632 (1983-11-11)
Nicarbazin, a drug used to control the protozoal disease coccidiosis in poultry, is a complex of the highly insoluble drug 4,4'-dinitrocarbanilide with 2-hydroxy-4,6-dimethylpyrimidine. The structures of this and other 4,4'-dinitrocarbanilide complexes have not been determined, but an analogous 2:1 complex
Randal S Stahl et al.
Journal of agricultural and food chemistry, 51(5), 1130-1135 (2003-02-20)
A method was developed using high-performance liquid chromatography to assay 4,4'-dinitrocarbanilide (DNC), the active ingredient in Nicarbazin, in eggshells collected from Canada geese fed a formulated feed fortified with Nicarbazin at doses of 0, 125, 250, and 500 microg/g. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service