388165
Titanium(IV) bis(ammonium lactato)dihydroxide solution
50 wt. % in H2O
Synonym(s):
Dihydroxybis(ammonium lactato)titanium solution
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
[CH3CH(O-)CO2NH4]2Ti(OH)2
CAS Number:
Molecular Weight:
294.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
0.51 psi ( 20 °C)
3.19 psi ( 55 °C)
Quality Level
form
liquid
reaction suitability
core: titanium
reagent type: catalyst
concentration
50 wt. % in H2O
refractive index
n20/D 1.438
density
1.222 g/mL at 25 °C
SMILES string
N.N.CC(O[Ti](O)(O)OC(C)C(O)=O)C(O)=O
InChI
1S/2C3H5O3.2H3N.2H2O.Ti/c2*1-2(4)3(5)6;;;;;/h2*2H,1H3,(H,5,6);2*1H3;2*1H2;/q2*-1;;;;;+4/p-2
InChI key
XRASGLNHKOPXQL-UHFFFAOYSA-L
Related Categories
Application
Titanium(IV) bis(ammonium lactato)dihydroxide can be used as:
- A water-soluble Ti precursor for the synthesis of TiO2/WO3 nanofibers applicable in photocatalysis.
- A reactant to synthesize a complex with pyrocatechol violet applicable as a colorimetric HF sensing dye.
- A Ti precursor to synthesize mesoporous silica loaded with titania nanoparticles for photocatalytic applications.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
81.9 °F
Flash Point(C)
27.7 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of TiO2/WO3 Composite Nanofibers by a Water-Based Electrospinning Process and Their Application in Photocatalysis
Odhiambo VO, et al.
Nanomaterials, 10(5), 882-882 (2020)
A pyrocatechol violet-titanium alkoxide complex for HF sensing: Study on the complex structure and application
Lee C, et al.
Dyes and Pigments, 127(5), 133-141 (2016)
Titania nanoparticle-loaded mesoporous silica synthesized through layer-by-layer assembly for the photodegradation of sodium dodecylbenzenesulfonate
Kim B, et al.
Applied Surface Science, 490(5), 38-46 (2019)
Preparation of colorimetric hydrogel beads for hydrofluoric acid detection
Lee C and Lee S-Y
Journal of Industrial and Engineering Chemistry, 38(5), 67-72 (2016)
Vincent Otieno Odhiambo et al.
Nanomaterials (Basel, Switzerland), 10(5) (2020-05-07)
TiO2/WO3 nanofibers were prepared in a one-step process by electrospinning. Titanium(IV) bis(ammonium lactato)dihydroxide (TiBALDH) and ammonium metatungstate (AMT) were used as water-soluble Ti and W precursors, respectively. Polyvinylpyrrolidone (PVP) and varying ratios of TiBALDH and AMT were dissolved in a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service