Skip to Content
Merck
All Photos(1)

Key Documents

376698

Sigma-Aldrich

(S)-(+)-4-Phenyl-2-oxazolidinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
Beilstein:
131918
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +48°, c = 2 in chloroform

mp

129-132 °C (lit.)

functional group

phenyl

SMILES string

O=C1N[C@H](CO1)c2ccccc2

InChI

1S/C9H9NO2/c11-9-10-8(6-12-9)7-4-2-1-3-5-7/h1-5,8H,6H2,(H,10,11)/t8-/m1/s1

InChI key

QDMNNMIOWVJVLY-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile chiral auxiliary for asymmetric synthesis which is easily recycled under mild conditions, thus enhancing its commercial potential. For a recent review, see Aldrichimica Acta .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ager, D.J., et al.
Aldrichimica Acta, 30, 3-3 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service