372102
4-Dodecylphenol, mixture of isomers
Synonym(s):
Dodecylphenol, n-Dodecylphenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C12H25C6H4OH
CAS Number:
Molecular Weight:
262.43
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
viscous liquid
Quality Level
refractive index
n20/D 1.503 (lit.)
bp
310-335 °C (lit.)
density
0.94 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCCc1ccc(O)cc1
InChI
1S/C18H30O/c1-2-3-4-5-6-7-8-9-10-11-12-17-13-15-18(19)16-14-17/h13-16,19H,2-12H2,1H3
InChI key
KJWMCPYEODZESQ-UHFFFAOYSA-N
Gene Information
rat ... Ar(24208)
General description
4-Dodecylphenol is an aromatic alcohol. Effect of exposure to 4-dodecylphenol was evaluated for the production of spleen cell specific cytokines IFN-γ and IL-4.
Application
4-Dodecylphenol is suitable for use in the preparation of polyaniline/4-dodecylphenol complex, an important organic corrosion inhibitor. It is suitable for use in the preparation of polymeric nanorods.
Modifier for the preparation of cycloaliphatic epoxy hybrids with amine curing agents
Catalyst for studies of thermal and thermomechanical behavior of novel cyanate ester homopolymers and copolymers
Catalyst for studies of thermal and thermomechanical behavior of novel cyanate ester homopolymers and copolymers
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effects of various chemicals including endocrine disruptors and analogs on the secretion of Th1 and Th2 cytokines from anti CD3-stimulated mouse spleen cells.
Yano K, et al.
Journal of Health Science, 49(3), 195-204 (2003)
Undoped polyaniline-surfactant complex for corrosion prevention.
Liu L-M and Levon K.
Journal of Applied Polymer Science, 73(14), 2849-2856 (1999)
Hollow Inorganic Nanospheres and Nanotubes with Tunable Wall Thicknesses by Atomic Layer Deposition on Self-Assembled Polymeric Templates.
Ras RHA, et al.
Advanced Materials, 19(1), 102-106 (2007)
Gabino Bolívar-Subirats et al.
Environmental science and pollution research international, 27(33), 41314-41325 (2020-07-18)
The aim of this study was to develop a high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS)-based method for the multiresidue analysis of 21 plastic additives in river water. Analysed compounds included phthalates, benzophenone, bisphenol A and long- and short-chain alkylphenols (APs)
Markus A Lill et al.
Journal of medicinal chemistry, 48(18), 5666-5674 (2005-09-02)
We investigated the influence of induced fit of the androgen receptor binding pocket on free energies of ligand binding. On the basis of a novel alignment procedure using flexible docking, molecular dynamics simulations, and linear-interaction energy analysis, we simulated the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service