Skip to Content
Merck
All Photos(2)

Key Documents

367494

Sigma-Aldrich

D-Glyceric acid calcium salt dihydrate

99%

Synonym(s):

(R)-2,3-Dihydroxypropanoic acid calcium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOCH2CH(OH)CO2]2Ca·2H2O
CAS Number:
Molecular Weight:
286.25
Beilstein:
6119062
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder or crystals

optical activity

[α]20/D +13°, c = 5 in H2O

mp

134 °C (dec.) (lit.)

SMILES string

O.O.OC[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)CO

InChI

1S/2C3H6O4.Ca.2H2O/c2*4-1-2(5)3(6)7;;;/h2*2,4-5H,1H2,(H,6,7);;2*1H2/q;;+2;;/p-2/t2*2-;;;/m11.../s1

InChI key

OKEVQKKVYQHNFW-AKYROZSLSA-L

Application

D-Glyceric acid calcium salt dihydrate can be used:
  • As an electrolyte for electrocatalytic oxidation of glycerol.
  • As a precursor to synthesize diacylester derivatives of glyceric acid esterified with palmitoyl chloride and linoleoyl chloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon nanotube supported PdAg nanoparticles for electrocatalytic oxidation of glycerol in anion exchange membrane fuel cells.
Benipal N, et al.
Applied Catalysis. B, Environmental, 210, 121-130 (2017)
Synthesis of dilinoleoyl-D-glyceric acid and evaluation of its cytotoxicity to human dermal fibroblast and endothelial cells.
Sato S, et al.
Journal of Oleo Science, 60(9), 483-487 (2011)
Stepwise synthesis of 2, 3-O-dipalmitoyl-D-glyceric acid and an in vitro evaluation of its cytotoxicity.
Shun S, et al.
Journal of Oleo Science, 61(6), 337-341 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service