363561
(R)-(−)-2-Chloro-1-phenylethanol
97%
Synonym(s):
(R)-(−)-α-(Chloromethyl)benzenemethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH(CH2Cl)OH
CAS Number:
Molecular Weight:
156.61
Beilstein:
4306747
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
optical activity
[α]20/D −48°, c = 2.8 in cyclohexane
refractive index
n20/D 1.552 (lit.)
density
1.185 g/mL at 25 °C (lit.)
functional group
chloro
hydroxyl
phenyl
SMILES string
O[C@@H](CCl)c1ccccc1
InChI
1S/C8H9ClO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H,6H2/t8-/m0/s1
InChI key
XWCQSILTDPAWDP-QMMMGPOBSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service