Skip to Content
Merck
All Photos(2)

Key Documents

362379

Sigma-Aldrich

Methyl trans-3-methoxyacrylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH=CHCO2CH3
CAS Number:
Molecular Weight:
116.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

56 °C/18 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

functional group

ester
ether

storage temp.

2-8°C

SMILES string

CO\C=C\C(=O)OC

InChI

1S/C5H8O3/c1-7-4-3-5(6)8-2/h3-4H,1-2H3/b4-3+

InChI key

AUTCCPQKLPMHDN-ONEGZZNKSA-N

Application

Methyl trans-3-methoxyacrylate may be used in the preparation of:
  • 5- hydroxyquinolines
  • 7-hydroxyquinolines
  • methyl 5-methoxyquinoline-3-carboxylate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Bohn et al.
Organic & biomolecular chemistry, 7(12), 2612-2618 (2009-06-09)
This work deals with the design of a bio-oxidisable prodrug strategy for the development of new central selective acetylcholinesterase inhibitors. This prodrug approach is expected to reduce peripheral anticholinesterase activity responsible for various side effects observed with presently marketed AChE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service