360058
(Triphenylsilyl)acetylene
98%
Synonym(s):
1,1′,1′′-(Ethynylsilylidyne)tris[benzene], Ethynyltriphenylsilane, Triphenylsilylacetylene, Triphenylsilylethyne
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
98%
mp
48-50 °C (lit.)
SMILES string
C#C[Si](c1ccccc1)(c2ccccc2)c3ccccc3
InChI
1S/C20H16Si/c1-2-21(18-12-6-3-7-13-18,19-14-8-4-9-15-19)20-16-10-5-11-17-20/h1,3-17H
InChI key
WADKYPSVXRWORK-UHFFFAOYSA-N
General description
(Triphenylsilyl)acetylene is a terminal alkyne. Rhodium-catalyzed asymmetric addition of (triphenylsilyl)acetylene to diphenylphosphinylallene is reported.
Application
(Triphenylsilyl)acetylene may be used in the synthesis of:
- selenothioic acid S-alkyl esters
- methyl 2-(di{ 3, 5-bis[(triphenylsilyl)ethynyl]phenyl}-phosphino)benzoate
- 2-(di{3,5-bis[(triphenylsilyl)ethynyl]phenyl}-phosphino)benzoic acid
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Crafting chiral space. The synthesis of C~ 2-symmetric diphosphine ligands for an outer-sphere catalytic reaction.
Bulletin de la Societe Chimique De France, 134, 263-274 (1997)
Selenothioic acid S-esters: Synthesis, characterization, and trend for stability.
Journal of the American Chemical Society, 119(37), 8592-8597 (1997)
Chemistry, an Asian journal, 3(8-9), 1505-1510 (2008-05-09)
The presence of an acid was found to be essential in the rhodium-catalyzed asymmetric addition of terminal alkynes to diarylphosphinylallenes giving exo-enynes in high yields with high regio- and enantioselectivity. The stereochemical outcome is determined at the protonolysis of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service