Skip to Content
Merck
All Photos(1)

Key Documents

359890

Sigma-Aldrich

Dimethyl bromomalonate

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH(COOCH3)2
CAS Number:
Molecular Weight:
211.01
Beilstein:
971135
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.460 (lit.)

bp

105-108 °C/11 mmHg (lit.)

density

1.601 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(Br)C(=O)OC

InChI

1S/C5H7BrO4/c1-9-4(7)3(6)5(8)10-2/h3H,1-2H3

InChI key

NEMOJKROKMMQBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl bromomalonate undergoes manganese(III)-promoted free-radical chain addition reaction with olefins, to yield dimethyl 2-bromoalkylmalonates. Organocatalyzed Michael addition of dimethyl bromomalonate to nitrostyrenes to yield synthetically useful nitrocyclopropanes has been reported. Dimethyl bromomalonate reacts readily with arylnitroso compounds to yield the corresponding N-aryl-C,C-dimethoxycarbonylnitrones.

Application

Dimethyl bromomalonate may be used in the enantioselective synthesis of nitrocyclopropanes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of functionalised cyclopropanes from nitroalkenes via an organocatalysed Michael-initiated ring-closure approach.
Russo A and Lattanzi A.
Tetrahedron Asymmetry, 21(9), 1155-1157 (2010)
Manganese (III)-promoted free-radical addition of dimethyl bromomalonate to olefins using an electrochemical regeneration procedure.
Nedelec JY and Nohair K.
Synlett, 09, 659-660 (1991)
Yi-Ning Xuan et al.
Organic letters, 11(7), 1583-1586 (2009-03-06)
Highly enantioselective synthesis of nitrocyclopropanes was achieved via the organocatalytic conjugate addition of dimethyl bromomalonate to nitroalkenes and the consequent intramolecular cyclopropanation. 6'-Demethyl quinine was found to be the efficient catalyst. Excellent enantioselectivities, diastereoselectivities, and good yields were obtained for
Synthesis and 1, 3-dipolar cycloaddition reactions of N-aryl-C, C-dimethoxycarbonylnitrones.
Tomioka Y, et al.
Journal of Heterocyclic Chemistry, 40(1), 121-128 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service