Skip to Content
Merck
All Photos(1)

Key Documents

35603

Sigma-Aldrich

(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine

≥97.0% (T)

Synonym(s):

(+)-3,3-Dichloro-2,N-epoxy-exo-10,2-bornanesultam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13Cl2NO3S
CAS Number:
Molecular Weight:
298.19
Beilstein:
3653352
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (T)

form

solid

optical activity

[α]20/D +89±3°, c = 0.5% in chloroform

functional group

chloro

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3(CS(=O)(=O)[N@@H]4O[C@@]34C1(Cl)Cl)C2(C)C

InChI

1S/C10H13Cl2NO3S/c1-7(2)6-3-4-8(7)5-17(14,15)13-10(8,16-13)9(6,11)12/h6H,3-5H2,1-2H3/t6-,8-,10+,13?/m0/s1

InChI key

HAMBQYFZDBYWHU-CNLAJYNUSA-N

Other Notes

Chiral oxidizing agent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F.A. Davis et al.
Tetrahedron Letters, 32, 7671-7671 (1991)
F.A. Davis et al.
The Journal of Organic Chemistry, 55, 3715-3715 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service