Skip to Content
Merck
All Photos(1)

Key Documents

347647

Sigma-Aldrich

5-Methyl-2-pyrazinecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
Beilstein:
116005
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

167-171 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1cnc(cn1)C(O)=O

InChI

1S/C6H6N2O2/c1-4-2-8-5(3-7-4)6(9)10/h2-3H,1H3,(H,9,10)

InChI key

RBYJWCRKFLGNDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methyl-2-pyrazinecarboxylic acid is a pyrazine derivative. Simultaneous membrane based solvent extraction (MBSE) and membrane based solvent stripping (MBSS) of 5-methyl-2-pyrazine carboxylic acid (MPCA) has been studied. Mechanism of pertraction of MPCA through layered bulk liquid membranes has been studied. MPCA reacts with Ln(NO3)3·6H2O and K4[W(CN)8]·2H2O to form unique three-dimensional network structure which contains open channels.

Application

5-Methyl-2-pyrazinecarboxylic acid may be used in the preparation of novel europium(III) complexes, (5-methyl-2-pyrazinecarboxylato) (phenanthroline)europium(III). It may be used in the synthesis of Ln-metal organic frameworks (MOFs).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile building-block synthesis of multifunctional lanthanide MOFs.
Tanase S, et al.
Journal of Materials Chemistry, 21(39), 15544-15551 (2011)
Transport of 5-methyl-2-pyrazinecarboxylic acid through a layered bulk liquid membrane.
Kubisova L, et al.
Chemical Papers, 56, 418-425 (2002)
Synthesis and photoluminescence properties of Eu(pzc)3(phen) and Eu(mpzc)3(phen).
Chang CH, et al.
Synthetic Metals, 145(1), 1-6 (2004)
Three-dimensional Ln III-W IV complexes with cyanido and carboxylato bridges. CrystEngComm 8.12 (2006): 863-865.
Tanase S, et al.
CrystEngComm, 8(12), 863-865 (2006)
Mass-transfer in membrane based solvent extraction and stripping of 5-methyl-2-pyrazinecarboxylic acid and co-transport of sulphuric acid in HF contactors.
Sabolova E, et al
Desalination, 163(1), 27-38 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service