Skip to Content
Merck
All Photos(3)

Key Documents

346802

Sigma-Aldrich

(S)-(−)-Indoline-2-carboxylic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

optical activity

[α]20/D −114°, c = 1 in 1 M HCl

mp

177 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)[C@@H]1Cc2ccccc2N1

InChI

1S/C9H9NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h1-4,8,10H,5H2,(H,11,12)/t8-/m0/s1

InChI key

QNRXNRGSOJZINA-QMMMGPOBSA-N

Application

Catalyst for enantioselective cyclopropanations.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roxanne K Kunz et al.
Journal of the American Chemical Society, 127(10), 3240-3241 (2005-03-10)
A new method for enantioselective organocatalytic cyclopropanation is described. This study outlines the identification of a new class of iminium catalyst based on the concept of directed electrostatic activation (DEA). This novel organocatalytic mechanism exploits dual activation of ylide and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service