Skip to Content
Merck
All Photos(1)

Key Documents

344419

Sigma-Aldrich

Copper(II) D-gluconate

98%

Synonym(s):

D-Gluconic acid copper(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22CuO14
CAS Number:
Molecular Weight:
453.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

optical activity

[α]26/D +15°, c = 1 in H2O

mp

155-157 °C (lit.)

application(s)

battery manufacturing

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Cu]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Cu/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;/h2*2-5,7-11H,1H2,(H,12,13);/q;;+2/p-2/t2*2-,3-,4+,5-;/m11./s1

InChI key

OCUCCJIRFHNWBP-IYEMJOQQSA-L

Application

Copper(II) D-gluconate powder in combination with silicon was used for the fabrication of carbon coated Si-Cu3Si composite as the anode materials for Li-ion batteries.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1B - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klára Szentmihályi et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 128, 110307-110307 (2020-06-01)
Despite significant nephrotoxicity, cisplatin is still used in the therapy of various tumors. We were interested in how metal ion composition is altered by cisplatin and whether platinum accumulates in the non-tumorous lung. We also aimed to study metal ion
Zhinan Li et al.
Comprehensive psychiatry, 102, 152200-152200 (2020-09-07)
Major depressive disorder (MDD) is frequently associated with cognitive deficits and high copper levels. Dysfunction of N-methyl-d-aspartate (NMDA) receptors has been postulated to underlie MDD pathogenesis. This study sought to investigate the curative effect of the NMDA receptor antagonist memantine
Robert A DiSilvestro et al.
Journal of the International Society of Sports Nutrition, 14, 42-42 (2017-11-22)
Certain essential and conditionally essential nutrients (CENs) perform functions involved in aerobic exercise performance. However, increased intake of such nutrient combinations has not actually been shown to improve such performance. For 1 mo, aerobically fit, young adult women took either
Robert A DiSilvestro et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 62, 126617-126617 (2020-07-13)
Elite female athletes have shown vulnerability to various degrees of iron deficiency. Less is known about recreational fitness exercisers. A study was done to examine plasma ferritin, an assessor of iron status, in young adult, university student fitness runners. The
Hong Hu et al.
Cancer medicine, 9(18), 6791-6801 (2020-08-05)
A growing number of studies show that intestinal microbiota affect the therapeutic effects of antineoplastic agents. Disulfiram (tetraethylthiuram disulfide, DSF) is an old alcohol-aversion drug that has been shown to be effective against various types of cancers in preclinical studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service