340871
4-Amino-5-chloro-2-methoxybenzoic acid
95%
Synonym(s):
4-Amino-5-chloro-o-anisic acid
About This Item
Quality Level
Assay
95%
form
solid
reaction suitability
reaction type: solution phase peptide synthesis
mp
206 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
COc1cc(N)c(Cl)cc1C(O)=O
InChI
1S/C8H8ClNO3/c1-13-7-3-6(10)5(9)2-4(7)8(11)12/h2-3H,10H2,1H3,(H,11,12)
InChI key
RVEATKYEARPWRE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service