Skip to Content
Merck
All Photos(3)

Key Documents

334162

Sigma-Aldrich

trans-1,2-Bis(phenylsulfonyl)ethylene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH=CHSO2C6H5
CAS Number:
Molecular Weight:
308.37
Beilstein:
2334889
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

221-223 °C (lit.)

functional group

sulfone

SMILES string

O=S(=O)(\C=C\S(=O)(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H12O4S2/c15-19(16,13-7-3-1-4-8-13)11-12-20(17,18)14-9-5-2-6-10-14/h1-12H/b12-11+

InChI key

YGBXMKGCEHIWMO-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chiral BINAP-AuTFA- and chiral BINAP-AgTFA (TFA = trifluoroacetate anion) -promoted catalytic enantioselective 1,3-dipolar cycloadditions of iminoglycinates with trans-1,2-bis(phenylsulfonyl)ethylene were studied.

Application

trans-1,2-Bis(phenylsulfonyl)ethylene was used in total synthesis of (+)-7-deoxypancratistatin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binap-gold (I) trifluoroacetate as a bifunctional catalyst for the synthesis of chiral prolines through 1, 3-dipolar cycloaddition of azomethine ylides.
Martin-Rodriguez M, et al.
Tetrahedron Asymmetry, 21(9), 1184-1186 (2010)
J L Aceña et al.
Organic letters, 2(23), 3683-3686 (2000-11-14)
A new total synthesis of (+)-7-deoxypancratistatin 1 has been accomplished in 19 steps (8% overall yield) from two readly available compounds, furan and trans-1,2-bis(phenylsulfonyl)ethylene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service