Skip to Content
Merck
All Photos(1)

Key Documents

331511

Sigma-Aldrich

1,2-Dichlorobenzene-d4

98 atom % D

Synonym(s):

Tetradeutero-1,2-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4Cl2
CAS Number:
Molecular Weight:
151.03
Beilstein:
1950096
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.5509 (lit.)

bp

178-180 °C (lit.)

mp

-17 °C (lit.)

density

1.341 g/mL at 25 °C (lit.)

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(Cl)c(Cl)c1[2H]

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H/i1D,2D,3D,4D

InChI key

RFFLAFLAYFXFSW-RHQRLBAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichlorobenzene-d4 is a deuterated NMR solvent that is useful in NMR-based research and analyses.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Eichelberger et al.
Journal of chromatographic science, 28(9), 460-467 (1990-09-01)
New drinking water regulations require the monitoring of eight volatile organic compounds that have established maximum contaminant levels (MCLs) and 51 other volatile organics for which MCLs are not established. A laboratory analytical method (Method 524.2) for the determination of
Magy. Kem. Foly., 98, 421-421 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service