Skip to Content
Merck
All Photos(2)

Key Documents

329584

Sigma-Aldrich

Zinc undecylenate

98%

Synonym(s):

Mycoseptin; Tineafax, 10-Undecylenic acid zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CH(CH2)8CO2]2Zn
CAS Number:
Molecular Weight:
431.92
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.22

vapor density

14.9 (vs air)

Quality Level

Assay

98%

form

powder

reaction suitability

core: zinc
reagent type: catalyst

mp

118-121 °C (lit.)

SMILES string

C=CCCCCCCCCC(=O)O[Zn]OC(=O)CCCCCCCCC=C

InChI

1S/2C11H20O2.Zn/c2*1-2-3-4-5-6-7-8-9-10-11(12)13;/h2*2H,1,3-10H2,(H,12,13);/q;;+2/p-2

InChI key

YMCOHQVWOBMDCZ-UHFFFAOYSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guncem Ozgun Eren et al.
STAR protocols, 2(3), 100664-100664 (2021-07-27)
Luminescent solar concentrators (LSCs) are simple and cost-effective solar energy-harvesting devices. Indium phosphide (InP)-based colloidal quantum dots (QDs) are promising QDs for efficient LSC devices due to their environmentally benign nature. One major challenge in LSC devices is reabsorption losses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service