Skip to Content
Merck
All Photos(1)

Key Documents

328294

Sigma-Aldrich

Dimethylphosphinic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2P(O)OH
CAS Number:
Molecular Weight:
94.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

85-89 °C (lit.)

SMILES string

CP(C)(O)=O

InChI

1S/C2H7O2P/c1-5(2,3)4/h1-2H3,(H,3,4)

InChI key

GOJNABIZVJCYFL-UHFFFAOYSA-N

Related Categories

General description

Infrared spectra for gaseous and polycrystalline samples of dimethylphosphinic acid at various temperatures are reported. The molecular structure of the hydrogen bonded cyclic dimer of dimethylphosphinic acid was studied by gas-phase electron diffraction at 433K.

Application

Dimethylphosphinic acid was used in preparation of microcrystalline organometallic coordination polymer.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas-phase electron diffraction study of cyclic dimer of dimethylphosphinic acid (Me< sub> 2</sub> P (? O) OH)< sub> 2</sub> using quantum chemical data and a priori force field.
Khaikin LS, et al.
Journal of Molecular Structure, 658(3), 153-170 (2003)
First example of the ABC ? (OH) absorption structure for both gaseous and crystalline phase: infrared studies of dimethylphosphinic acid.
Tokhadze KG, et al.
Journal of Molecular Structure, 404(1), 55-62 (1997)
Christopher M. Barnes et al.
Inorganic chemistry, 36(25), 5793-5798 (2001-10-24)
The microcrystalline organometallic coordination polymer [Ru(2){&mgr;(2):&mgr;(2):eta(2)-O(2)PMe(2)}(2)(CO)(4)](n)() which results from the oxidative addition of dimethylphosphinic acid to triruthenium dodecacarbonyl has been structurally characterized by X-ray powder diffraction, lambda = 1.149 49(1) Å, at 295 and 50 K. At room temperature the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service