Skip to Content
Merck

326356

Sigma-Aldrich

Niobium(III) chloride 1,2-dimethoxyethane complex

Synonym(s):

Niobium trichloride 1,2-dimethoxyethane complex, Trichloro(1,2-dimethoxyethane)niobium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NbCl3 · CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
289.39
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

form

powder

Quality Level

concentration

30.6-33.5% (%Nb after ignition to oxide, gravimetric)

mp

230 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[Nb](Cl)Cl.COCCOC

InChI

1S/C4H10O2.3ClH.Nb/c1-5-3-4-6-2;;;;/h3-4H2,1-2H3;3*1H;/q;;;;+3/p-3

InChI key

SBSZUBUHOUKQMN-UHFFFAOYSA-K

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
357405464171339237
assay

99.9% trace metals basis

assay

≥99.9% trace metals basis

assay

97%

assay

99.9% trace metals basis

density

4.43 g/mL (lit.)

density

2.8 g/mL at 25 °C (lit.)

density

3.25 g/mL at 25 °C (lit.)

density

-

impurities

≤1500.0 ppm Trace Metal Analysis

impurities

≤1000.0 ppm Trace Metal Analysis

impurities

-

impurities

-

mp

910 °C (lit.)

mp

-

mp

-

mp

-

reaction suitability

reagent type: catalyst
core: zirconium

reaction suitability

reagent type: catalyst
core: zirconium

reaction suitability

-

reaction suitability

-

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service