Skip to Content
Merck

326275

Sigma-Aldrich

2,2,3,3-Tetrafluoro-1-propanol

98%

Synonym(s):

α,α,ψ-Trihydroperfluoropropanol, 1,1,2,2-Tetrafluoro-3-hydroxypropane, 1,1,3-Trihydroperfluoro-1-propanol, 1-Hydroxy-2,2,3,3-tetrafluoropropane, 1H,1H,3H-Perfluoropropanol, 1H,1H,3H-Tetrafluoro-1-propanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€260.00

€260.00


Availability
Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€260.00

About This Item

Linear Formula:
CHF2CF2CH2OH
CAS Number:
Molecular Weight:
132.06
Beilstein:
1738264
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€260.00


Availability
Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.321 (lit.)

bp

107-109 °C (lit.)

density

1.48 g/mL at 20 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

OCC(F)(F)C(F)F

InChI

1S/C3H4F4O/c4-2(5)3(6,7)1-8/h2,8H,1H2

InChI key

NBUKAOOFKZFCGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,3,3-Tetrafluoro-1-propanol (TFP) is extensively used as the solvent.[1] Mineralization and defluoridation of TFP by UV oxidation in a novel three-phase fluidized bed reactor was reported.[1] Desorption of activated carbon loaded with TFP by supercritical carbon dioxide in a rotating packed bed was investigated.[2]

Application


  • Water repellent applications: 2,2,3,3-Tetrafluoro-1-propanol is utilized in the synthesis of vegetable oil-based fluorocarbon water repellent agents for cotton fabrics, demonstrating its utility in enhancing the water resistance of textiles, important for clothing and outdoor gear industries (Zhao et al., 2012).

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Jen Shih et al.
Water research, 47(7), 2325-2330 (2013-03-05)
2,2,3,3-Tetrafluoro-1-propanol (TFP, C3H4F4O, M.W. = 132.06) is extensively used as the solvent in CD-R and DVD-R fabrication. Since it has a fluorinated alky-chain configuration and is non-biodegradable, its treatment by conventional oxidation methods is typically very inefficient. In this work
Chung-Sung Tan et al.
Environmental science & technology, 42(6), 2150-2154 (2008-04-16)
Desorption of activated carbon loaded with 2,2,3,3-tetrafluoro-1-propanol (TFP) by supercritical carbon dioxide in a rotating packed bed was investigated in this study. The experimental data show that the time required to achieve complete desorption of TFP from activated carbon in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service