Skip to Content
Merck
All Photos(1)

Key Documents

317748

Sigma-Aldrich

N-Allylmethylamine

96%

Synonym(s):

N-Methylallylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2NHCH3
CAS Number:
Molecular Weight:
71.12
Beilstein:
1098387
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

64-66 °C (lit.)

density

0.741 g/mL at 25 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CNCC=C

InChI

1S/C4H9N/c1-3-4-5-2/h3,5H,1,4H2,2H3

InChI key

IOXXVNYDGIXMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hannah C Bygd et al.
Biomaterials, 56, 187-197 (2015-05-03)
Macrophage reprogramming has long been the focus of research in disease therapeutics and biomaterial implantation. With different chemical and physical properties of materials playing a role in macrophage polarization, it is important to investigate and categorize the activation effects of
Daniel Wang et al.
Biomacromolecules, 15(11), 4102-4110 (2014-10-01)
Material properties play a key role in the cellular internalization of polymeric particles. In the present study, we have investigated the effects of material characteristics such as water contact angle, zeta potential, melting temperature, and alternative activation of complement on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service