Skip to Content
Merck
All Photos(2)

Key Documents

317691

Sigma-Aldrich

Allyl disulfide

technical grade, 80%

Synonym(s):

Diallyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2SSCH2CH=CH2
CAS Number:
Molecular Weight:
146.27
Beilstein:
1699241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor density

>5 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

80%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

180-195 °C (lit.)

density

1.008 g/mL at 25 °C (lit.)

functional group

disulfide

storage temp.

2-8°C

SMILES string

C=CCSSCC=C

InChI

1S/C6H10S2/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-6H2

InChI key

PFRGXCVKLLPLIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Organosulfur compound that is naturally found in garlic. Contributes to the pungent odor of crushed garlic. Anticancer agent and may provide protection against cardiovascular disease.

Other Notes

Remainder other allyl sulfides

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel Edward Bennett Saidu et al.
Bioorganic & medicinal chemistry, 20(4), 1584-1593 (2012-01-24)
Coumarins and coumarin derivatives as well as diallyl polysulfides are well known as anticancer drugs. In order to find new drugs with anticancer activities, we combined coumarins with polysulfides in the form of di-coumarin polysulfides. These novel compounds were tested
Tao Zeng et al.
Biochimica et biophysica acta, 1830(10), 4848-4859 (2013-07-03)
Diallyl disulfide (DADS) is a garlic-derived organosulfur compound. The current study is designed to evaluate the protective effects of DADS against ethanol-induced oxidative stress, and to explore the underlying mechanisms by examining the HO-1/Nrf-2 pathway. We investigated whether or not
Laura Masuelli et al.
Frontiers in bioscience (Landmark edition), 17, 498-508 (2011-12-29)
Malignant tumors of mesenchimal origin such as rhabdomyosarcoma and osteosarcoma are highly aggressive pedriatic malignancies with a poor prognosis. Indeed, the initial response to chemotherapy is followed by chemoresistance. Diallyl disulfide (DADS), resveratrol (RES) and curcumin (CUR) are dietary chemopreventive
Tatsuya Sueyoshi et al.
PloS one, 6(6), e21229-e21229 (2011-06-24)
Constituent chemicals in garlic extract are known to induce phase I and phase II enzymes in rodent livers. Here we have utilized Car(+/+) and Car(-/-) mice to demonstrate that the nuclear xenobiotic receptor CAR regulated the induction of the estrogen
Akeem O Lawal et al.
Environmental toxicology and pharmacology, 32(2), 266-274 (2011-08-17)
Garlic has been reported in many previous studies as a potent chemopreventive agent. The protective effect of garlic has been ascribed to the presence of organosulphur compounds (OSC). In this study, the efficacy of aged garlic extract (AGE) compared to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service