Skip to Content
Merck
All Photos(4)

Key Documents

316385

Sigma-Aldrich

Potassium fluoride on alumina

greener alternative

extent of labeling: 40 wt. % loading

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

40 wt. % loading

greener alternative category

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Green etherification reagent of choice according to Glaxo-Smith-Kline's reagent guide.

Synthesis of diaryl ethers, diaryl thioethers, and diarylamines mediated by potassium fluoride-alumina and 18-crown-6
Potassium fluoride loaded alumina can be used to catalyze:
  • Dehydration of alkyl and aryl aldoximes into nitriles may be catalyze by potassium fluoride loaded on alumina.
  • Palladium-catalysed reactions to prepare allylic thiol derivatives.
  • N-Alkylation of carboxamides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium fluoride doped on alumina: An efficient catalyst for conversion of aldoximes into nitriles.
Movassagh B and Shokri S
Synthetic Communications, 35(6), 887-890 (2005)
Microwave mediated palladium-catalysed reactions on potassium fluoride/alumina without use of solvent.
Villemin D and Caillot F
Tetrahedron Letters, 42(4), 639-642 (2001)
N-Alkylation of amides and N-heterocycles with potassium fluoride on alumina.
Yamawaki J, et al.
Chemistry Letters (Jpn), 10(8), 1143-1146 (1981)
Deniz Ekinci et al.
Bioorganic & medicinal chemistry, 19(2), 744-748 (2011-01-08)
Carbonic anhydrase (CA; EC 4.2.1.1) was purified from the gill of the teleost fish Dicentrarchus labrax (European seabass). The purification procedure consisted of a single step affinity chromatography on Sepharose 4B-tyrosine-sulfanilamide. The enzyme was purified 84.9-fold with a yield of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service