Skip to Content
Merck
All Photos(2)

Key Documents

309346

Sigma-Aldrich

L-Glutamic acid diethyl ester hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH2CH2CH(NH2)COOC2H5 · HCl
CAS Number:
Molecular Weight:
239.70
Beilstein:
3597595
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]18/D +22°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

108-110 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].CCOC(=O)CC[C@H](N)C(=O)OCC

InChI

1S/C9H17NO4.ClH/c1-3-13-8(11)6-5-7(10)9(12)14-4-2;/h7H,3-6,10H2,1-2H3;1H/t7-;/m0./s1

InChI key

WSEQLMQNPBNMSL-FJXQXJEOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Glutamic acid diethyl ester hydrochloride can be used in the synthesis of:
  • Oligo(γ-ethyl L-glutamate) via oligomerization catalyzed by papain.
  • L-glutamic acid based dendritic compounds.
  • Poly(α-peptide) by polymerization and copolymerization in the presence of protease catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound induced formation of organogel from a glutamic dendron
Li Y, et al.
Tetrahedron, 63(31), 7468-7473 (2007)
M G Matheus et al.
Psychopharmacology, 132(1), 14-18 (1997-07-01)
Microinjections of glutamate into the dorsal periaqueductal grey (DPAG) of rats induce flight behavior, and blockade of glutamate NMDA receptors in the same region increases exploratory behavior of rats tested on the elevated plus maze. To investigate a possible role
Rapid regioselective oligomerization of L-glutamic acid diethyl ester catalyzed by papain
Li G, et al.
Macromolecules, 39(23), 7915-7921 (2006)
Inhibition of cocaine intoxication by excitatory amino acid receptor antagonists.
R W Rockhold et al.
Annals of the New York Academy of Sciences, 648, 335-337 (1992-05-11)
P D Evans et al.
The Journal of experimental biology, 173, 229-249 (1992-12-01)
L-Glutamate application can produce three different responses in the membrane potential of the Schwann cell of the tropical squid, Sepioteuthis sepioidea, which appear to be mediated by three pharmacologically distinct classes of receptor. A class of non-NMDA-type receptors, with some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service