Skip to Content
Merck
All Photos(1)

Key Documents

307084

Sigma-Aldrich

(R)-(−)-2-Amino-1-butanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1718929
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]19/D −10°, neat

optical purity

ee: 96% (GLC)

refractive index

n20/D 1.452

bp

172-174 °C (lit.)

mp

−2 °C (lit.)

density

0.943 g/mL at 20 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CC[C@@H](N)CO

InChI

1S/C4H11NO/c1-2-4(5)3-6/h4,6H,2-3,5H2,1H3/t4-/m1/s1

InChI key

JCBPETKZIGVZRE-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Datong Wu et al.
The Analyst, 145(9), 3395-3400 (2020-04-03)
Here, ionized chiral carbon dots, (S,S)-C-dots-1 (λex = 430 nm, λem = 480 nm), were synthesized via a facile route with relatively high quantum yield (∼24.4%) and used as a fluorescent chiral sensor. One of the advantages of the synthetic
Rym Hassani et al.
Chemico-biological interactions, 217, 41-48 (2014-04-15)
This paper describes the synthesis of new enantiomerically pure 2-cyanoethyl-oxazolines in one step starting from a wide range of amino alcohols and 4-ethoxy-4-iminobutanenitrile with high to good yields (73-96%) via an appropriate procedure which can be used for a selective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service