Skip to Content
Merck
All Photos(2)

Key Documents

306665

Sigma-Aldrich

Dodecanedioyl dichloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO(CH2)10COCl
CAS Number:
Molecular Weight:
267.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.75 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.468 (lit.)

bp

140 °C/0.5 mmHg (lit.)

density

1.069 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)CCCCCCCCCCC(Cl)=O

InChI

1S/C12H20Cl2O2/c13-11(15)9-7-5-3-1-2-4-6-8-10-12(14)16/h1-10H2

InChI key

CNXXEPWXNDFGIG-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Queeny Dasgupta et al.
Physical chemistry chemical physics : PCCP, 18(3), 2112-2119 (2015-12-23)
Salicylic acid (SA) based biodegradable polyanhydrides (PAHs) are of great interest for drug delivery in a variety of diseases and disorders owing to the multi-utility of SA. There is a need for the design of SA-based PAHs for tunable drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service