Skip to Content
Merck
All Photos(2)

Key Documents

305863

Sigma-Aldrich

2-(3-Butynyloxy)tetrahydro-2H-pyran

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O2
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.457 (lit.)

bp

92-95 °C/18 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

C#CCCOC1CCCCO1

InChI

1S/C9H14O2/c1-2-3-7-10-9-6-4-5-8-11-9/h1,9H,3-8H2

InChI key

ZQZSNKJFOFAJQX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guy Bertrand Djigoué et al.
Molecules (Basel, Switzerland), 18(1), 914-933 (2013-01-25)
We synthesized two series of androstane derivatives as inhibitors of type 3 and type 5 17β-hydroxysteroid dehydrogenases (17β-HSDs). In the first series, four monospiro derivatives at position C17 were prepared from androsterone (ADT) or epi-ADT. After the protection of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service